Assay Details
| Gene Symbol: BRMS1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261019.1 | NP_001247948.1 | 781 | 103 | ||
| XM_028850211.1 | XP_028706044.1 | 717 | 103 | |||
| GenBank mRNA | JU336709.1 | - | 801 | 103 | ||
| JV048441.1 | - | 781 | 103 | |||
More Information
Set Membership: |
|

BRMS1L
Genomic Map