Assay Details
| Gene Symbol: TRMT11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258056.1 | NP_001244985.1 | 1275 | 89 | ||
| XM_028846802.1 | XP_028702635.1 | 1182 | 89 | |||
| GenBank mRNA | JU346784.1 | - | 725 | 89 | ||
| JU476678.1 | - | 1275 | 89 | |||
More Information
Set Membership: |
|

TRMT11
Genomic Map