Assay Details
Gene Symbol: WASHC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015152599.2 | XP_015008085.1 | 277 | 118 | ||
XM_015152600.2 | XP_015008086.1 | 277 | 118 | |||
XM_015152601.2 | XP_015008087.1 | 277 | 118 | |||
XM_015152602.2 | XP_015008088.1 | 306 | 118 | |||
XM_015152603.2 | XP_015008089.1 | 306 | 118 | |||
XM_015152605.1 | XP_015008091.1 | 277 | 118 | |||
XM_015152606.2 | XP_015008092.1 | 306 | 118 | |||
XM_015152607.2 | XP_015008093.1 | 277 | 118 | |||
XM_028829811.1 | XP_028685644.1 | 277 | 118 | |||
GenBank mRNA | JU334760.1 | - | 244 | 118 | ||
JU473164.1 | - | 237 | 118 | |||
JV043805.1 | - | 249 | 118 | |||
JV453124.1 | - | 215 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() |
