Assay Details
| Gene Symbol: SWAP70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172414.2 | NP_001165885.1 | 1000 | 140 | ||
| XM_015114742.2 | XP_014970228.1 | 1005 | 140 | |||
| XM_028832607.1 | XP_028688440.1 | 1009 | 140 | |||
| GenBank mRNA | JU322000.1 | - | 931 | 140 | ||
| JU473817.1 | - | 979 | 140 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SWAP70
Genomic Map