Assay Details
| Gene Symbol: IFIT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028826634.1 | XP_028682467.1 | 350 | 88 | ||
| XM_028826635.1 | XP_028682468.1 | 95 | 88 | |||
| GenBank mRNA | JU334337.1 | - | 105 | 88 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028826634.1, XM_028826635.1 transcripts.
Set Membership: |
|

IFIT1
Genomic Map