Assay Details
| Gene Symbol: HAND2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193765.2 | NP_001180694.1 | 461 | 63 | ||
| XM_001085842.4 | XP_001085842.1 | 1493 | 63 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HAND2| Gene Symbol: HAND2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193765.2 | NP_001180694.1 | 461 | 63 | ||
| XM_001085842.4 | XP_001085842.1 | 1493 | 63 | |||
Set Membership: |
|