Assay Details
Gene Symbol: LOC695267 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015146698.2 | XP_015002184.2 | 6232 | 64 | ||
XM_015146699.2 | XP_015002185.2 | 5836 | 64 | |||
XM_015146702.2 | XP_015002188.2 | 5774 | 64 | |||
XM_015146704.2 | XP_015002190.2 | 5803 | 64 | |||
XM_015146705.2 | XP_015002191.2 | 6154 | 64 | |||
XM_015146706.2 | XP_015002192.2 | 6136 | 64 | |||
XM_015146707.2 | XP_015002193.2 | 6106 | 64 | |||
XM_015146708.2 | XP_015002194.2 | 6061 | 64 | |||
XM_015146709.2 | XP_015002195.2 | 6010 | 64 | |||
XM_015146710.2 | XP_015002196.2 | 5308 | 64 | |||
XM_015146711.2 | XP_015002197.2 | 5137 | 64 | |||
XM_015146713.2 | XP_015002199.2 | 5056 | 64 | |||
XM_015146714.2 | XP_015002200.2 | 4960 | 64 | |||
XM_015146715.2 | XP_015002201.2 | 4834 | 64 | |||
XM_028826127.1 | XP_028681960.1 | 5969 | 64 | |||
XM_028826128.1 | XP_028681961.1 | 5182 | 64 | |||
XM_028826130.1 | XP_028681963.1 | 4930 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
