Assay Details
| Gene Symbol: FBXO3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257701.1 | NP_001244630.1 | 696 | 78 | ||
| XM_015114494.2 | XP_014969980.1 | 722 | 78 | |||
| XM_015114495.2 | XP_014969981.1 | 722 | 78 | |||
| GenBank mRNA | JU333949.1 | - | 696 | 78 | ||
| JU475133.1 | - | 690 | 78 | |||
| JV043949.1 | - | 706 | 78 | |||
| JV648100.1 | - | 706 | 78 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

FBXO3
Genomic Map