Assay Details
| Gene Symbol: PSMA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260609.2 | NP_001247538.1 | 371 | 95 | ||
| XM_015146783.2 | XP_015002269.1 | 333 | 95 | |||
| GenBank mRNA | JU335513.1 | - | 366 | 95 | ||
| JU476453.1 | - | 372 | 95 | |||
| JV047692.1 | - | 371 | 95 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PSMA5
Genomic Map