Assay Details
| Gene Symbol: ZSCAN18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015125018.2 | XP_014980504.2 | 1029 | 66 | ||
| XM_015125020.2 | XP_014980506.2 | 1017 | 66 | |||
| XM_015125022.2 | XP_014980508.2 | 1029 | 66 | |||
| XM_015125023.2 | XP_014980509.2 | 1020 | 66 | |||
| XM_028838793.1 | XP_028694626.1 | 851 | 66 | |||
| XM_028838794.1 | XP_028694627.1 | 1118 | 66 | |||
| GenBank mRNA | JU347518.1 | - | 879 | 66 | ||
More Information
Set Membership: |
|

ZSCAN18
Genomic Map