Assay Details
| Gene Symbol: PLSCR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265888.1 | NP_001252817.1 | 824 | 56 | ||
| XM_015118456.2 | XP_014973942.1 | 1214 | 56 | |||
| XM_015118457.2 | XP_014973943.1 | 1218 | 56 | |||
| XM_028835783.1 | XP_028691616.1 | 1309 | 56 | |||
| GenBank mRNA | JU321589.1 | - | 828 | 56 | ||
| JV045213.1 | - | 824 | 56 | |||
More Information
Set Membership: |
|

PLSCR3
Genomic Map