Assay Details
| Gene Symbol: CSRP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001095430.4 | XP_001095430.1 | 410 | 56 | ||
| XM_015114651.2 | XP_014970137.1 | 326 | 56 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CSRP3| Gene Symbol: CSRP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001095430.4 | XP_001095430.1 | 410 | 56 | ||
| XM_015114651.2 | XP_014970137.1 | 326 | 56 | |||
Set Membership: |
|