Assay Details
| Gene Symbol: EFNB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261375.1 | NP_001248304.1 | 802 | 60 | ||
| GenBank mRNA | JU474587.1 | - | 897 | 60 | ||
| JV043851.1 | - | 802 | 60 | |||
More Information
Set Membership: |
|

EFNB1
Genomic Map