Assay Details
| Gene Symbol: DNAAF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002805231.3 | XP_002805277.3 | 3212 | 91 | ||
| GenBank mRNA | JU325030.1 | - | 683 | 91 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DNAAF2| Gene Symbol: DNAAF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002805231.3 | XP_002805277.3 | 3212 | 91 | ||
| GenBank mRNA | JU325030.1 | - | 683 | 91 | ||
Set Membership: |
|