Assay Details
| Gene Symbol: TSN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266679.1 | NP_001253608.1 | 285 | 65 | ||
| XM_015109890.2 | XP_014965376.1 | 1000 | 65 | |||
| GenBank mRNA | JU336175.1 | - | 378 | 65 | ||
| JU477032.1 | - | 440 | 65 | |||
| JV048126.1 | - | 285 | 65 | |||
| JV048127.1 | - | 285 | 65 | |||
More Information
Set Membership: |
|

TSN
Genomic Map