Assay Details
| Gene Symbol: VANGL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015113503.2 | XP_014968989.1 | 1428 | 60 | ||
| XM_015113509.2 | XP_014968995.1 | 3247 | 60 | |||
| XM_015113518.2 | XP_014969004.1 | 2811 | 60 | |||
| XM_015113519.2 | XP_014969005.1 | 1537 | 60 | |||
| XM_015113524.2 | XP_014969010.1 | 1512 | 60 | |||
| XM_028827040.1 | XP_028682873.1 | 1453 | 60 | |||
| GenBank mRNA | JU335599.1 | - | 1429 | 60 | ||
| JU476511.1 | - | 1132 | 60 | |||
More Information
Set Membership: |
|

VANGL2
Genomic Map