Assay Details
| Gene Symbol: RCOR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028851874.1 | XP_028707707.1 | 584 | 73 | ||
| XM_028851875.1 | XP_028707708.1 | 555 | 73 | |||
| XM_028851876.1 | XP_028707709.1 | 596 | 73 | |||
| GenBank mRNA | JV055795.1 | - | 144 | 73 | ||
| JV710358.1 | - | 166 | 73 | |||
More Information
Set Membership: |
|

RCOR1
Genomic Map