Assay Details
| Gene Symbol: FBXO31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258079.1 | NP_001245008.1 | 685 | 75 | ||
| XM_015126750.2 | XP_014982236.1 | 810 | 75 | |||
| XM_028840364.1 | XP_028696197.1 | 810 | 75 | |||
| XM_028840365.1 | XP_028696198.1 | 810 | 75 | |||
| XM_028840367.1 | XP_028696200.1 | 651 | 75 | |||
| XM_028840368.1 | XP_028696201.1 | 810 | 75 | |||
| XM_028840369.1 | XP_028696202.1 | 723 | 75 | |||
| GenBank mRNA | JU477237.1 | - | 685 | 75 | ||
More Information
Set Membership: |
|

FBXO31
Genomic Map