Assay Details
| Gene Symbol: NSDHL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265710.1 | NP_001252639.1 | 534 | 75 | ||
| XM_015128441.2 | XP_014983927.1 | 673 | 75 | |||
| XM_015128444.2 | XP_014983930.1 | 477 | 75 | |||
| XM_028841645.1 | XP_028697478.1 | 787 | 75 | |||
| XM_028841646.1 | XP_028697479.1 | 569 | 75 | |||
| XM_028841647.1 | XP_028697480.1 | 599 | 75 | |||
| GenBank mRNA | JU323097.1 | - | 584 | 75 | ||
| JU475789.1 | - | 559 | 75 | |||
| JV045287.1 | - | 558 | 75 | |||
| JV667235.1 | - | 534 | 75 | |||
More Information
Set Membership: |
|

NSDHL
Genomic Map