Assay Details
| Gene Symbol: MYZAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028850979.1 | XP_028706812.1 | 1142 | 107 | ||
| XM_028850980.1 | XP_028706813.1 | 1142 | 107 | |||
| GenBank mRNA | JU321531.1 | - | 1125 | 107 | ||
| JV704273.1 | - | 1132 | 107 | |||
More Information
Set Membership: |
|

MYZAP
Genomic Map