Assay Details
| Gene Symbol: CDH9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015139773.2 | XP_014995259.1 | 1061 | 121 | ||
| GenBank mRNA | JU321902.1 | - | 802 | 121 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CDH9| Gene Symbol: CDH9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015139773.2 | XP_014995259.1 | 1061 | 121 | ||
| GenBank mRNA | JU321902.1 | - | 802 | 121 | ||
Set Membership: |
|