Assay Details
| Gene Symbol: ORC6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194605.2 | NP_001181534.1 | 518 | 78 | ||
| XM_015126044.2 | XP_014981530.1 | 841 | 78 | |||
| XM_015126046.2 | XP_014981532.1 | 910 | 78 | |||
| XM_028840582.1 | XP_028696415.1 | 1330 | 78 | |||
| GenBank mRNA | JU474980.1 | - | 495 | 78 | ||
| JV710982.1 | - | 491 | 78 | |||
More Information
Set Membership: |
|

ORC6
Genomic Map