Assay Details
| Gene Symbol: FYN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015137565.2 | XP_014993051.1 | 2549 | 91 | ||
| XM_028847044.1 | XP_028702877.1 | 2012 | 91 | |||
| XM_028847045.1 | XP_028702878.1 | 1563 | 91 | |||
| XM_028847046.1 | XP_028702879.1 | 1645 | 91 | |||
| XM_028847047.1 | XP_028702880.1 | 1489 | 91 | |||
| GenBank mRNA | JU319770.1 | - | 1635 | 91 | ||
| JU333777.1 | - | 1626 | 91 | |||
| JU475022.1 | - | 1991 | 91 | |||
| JU475023.1 | - | 1982 | 91 | |||
| JU475024.1 | - | 1826 | 91 | |||
| JV046640.1 | - | 1667 | 91 | |||
| JV451748.1 | - | 1444 | 91 | |||
| JV647292.1 | - | 1991 | 91 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

FYN
Genomic Map