Assay Details
| Gene Symbol: MYBL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265855.1 | NP_001252784.1 | 747 | 64 | ||
| GenBank mRNA | JU475685.1 | - | 747 | 64 | ||
| JV652126.1 | - | 853 | 64 | |||
More Information
Set Membership: |
|

MYBL2
Genomic Map