Assay Details
| Gene Symbol: HIVEP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028847169.1 | XP_028703002.1 | 7210 | 87 | ||
| XM_028847170.1 | XP_028703003.1 | 5995 | 87 | |||
| XM_028847171.1 | XP_028703004.1 | 6255 | 87 | |||
| XM_028847172.1 | XP_028703005.1 | 10131 | 87 | |||
| XM_028847173.1 | XP_028703006.1 | 7142 | 87 | |||
| XM_028847174.1 | XP_028703007.1 | 7264 | 87 | |||
| XM_028847175.1 | XP_028703008.1 | 5928 | 87 | |||
| GenBank mRNA | JU470872.1 | - | 5852 | 87 | ||
| JV703519.1 | - | 5846 | 87 | |||
More Information
Set Membership: |
|

HIVEP2
Genomic Map