Assay Details
| Gene Symbol: FRMD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015143686.2 | XP_014999172.1 | 1140 | 67 | ||
| XM_015143687.2 | XP_014999173.1 | 1191 | 67 | |||
| XM_015143688.2 | XP_014999174.1 | 1049 | 67 | |||
| XM_015143689.2 | XP_014999175.1 | 1116 | 67 | |||
| XM_028851548.1 | XP_028707381.1 | 1676 | 67 | |||
| XM_028851549.1 | XP_028707382.1 | 1244 | 67 | |||
| GenBank mRNA | JU321839.1 | - | 1116 | 67 | ||
| JV044054.1 | - | 1107 | 67 | |||
More Information
Set Membership: |
|

FRMD6
Genomic Map