Assay Details
| Gene Symbol: ZC3HAV1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001107704.3 | XP_001107704.1 | 784 | 74 | ||
| GenBank mRNA | JU496263.1 | - | 367 | 74 | ||
| JV075521.1 | - | 60 | 74 | |||
More Information
Set Membership: |
|

ZC3HAV1L
Genomic Map