Assay Details
| Gene Symbol: TEK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001105270.4 | XP_001105270.2 | 640 | 66 | ||
| XM_015117580.2 | XP_014973066.2 | 640 | 66 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TEK| Gene Symbol: TEK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001105270.4 | XP_001105270.2 | 640 | 66 | ||
| XM_015117580.2 | XP_014973066.2 | 640 | 66 | |||
Set Membership: |
|