Assay Details
| Gene Symbol: POLG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015120114.2 | XP_014975600.2 | 668 | 137 | ||
| GenBank mRNA | JU324087.1 | - | 640 | 137 | ||
| JV706008.1 | - | 637 | 137 | |||
More Information
Set Membership: |
|

POLG2
Genomic Map