Assay Details
| Gene Symbol: CARNMT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001094584.4 | XP_001094584.2 | 663 | 138 | ||
| XM_028835250.1 | XP_028691083.1 | 765 | 138 | |||
| XM_028835251.1 | XP_028691084.1 | 563 | 138 | |||
| XM_028835252.1 | XP_028691085.1 | 664 | 138 | |||
| GenBank mRNA | JU327736.1 | - | 278 | 138 | ||
| JV635966.1 | - | 579 | 138 | |||
| JV707245.1 | - | 304 | 138 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CARNMT1
Genomic Map