Assay Details
| Gene Symbol: HCLS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257947.1 | NP_001244876.1 | 405 | 66 | ||
| XM_015129912.2 | XP_014985398.2 | 394 | 66 | |||
| GenBank mRNA | JU334240.1 | - | 405 | 66 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

HCLS1
Genomic Map