Assay Details
| Gene Symbol: BUB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015148313.2 | XP_015003799.1 | 1212 | 134 | ||
| GenBank mRNA | JU477806.1 | - | 1192 | 134 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

BUB3| Gene Symbol: BUB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015148313.2 | XP_015003799.1 | 1212 | 134 | ||
| GenBank mRNA | JU477806.1 | - | 1192 | 134 | ||
Set Membership: |
|