Assay Details
| Gene Symbol: GDPD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194431.2 | NP_001181360.1 | 792 | 59 | ||
| GenBank mRNA | JU493309.1 | - | 560 | 59 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GDPD3| Gene Symbol: GDPD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194431.2 | NP_001181360.1 | 792 | 59 | ||
| GenBank mRNA | JU493309.1 | - | 560 | 59 | ||
Set Membership: |
|