Assay Details
| Gene Symbol: PPP2R5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257639.1 | NP_001244568.1 | 1804 | 97 | ||
| XM_015116924.2 | XP_014972410.1 | 1219 | 97 | |||
| XM_015116930.2 | XP_014972416.1 | 1280 | 97 | |||
| GenBank mRNA | JU335286.1 | - | 1804 | 97 | ||
| JU476242.1 | - | 1739 | 97 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PPP2R5A
Genomic Map