Assay Details
| Gene Symbol: CTSB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194899.2 | NP_001181828.2 | 1030 | 81 | ||
| XM_015144727.2 | XP_015000213.2 | 1109 | 81 | |||
| XM_015144728.2 | XP_015000214.2 | 963 | 81 | |||
| XM_015144736.2 | XP_015000222.1 | 550 | 81 | |||
| XM_028852097.1 | XP_028707930.1 | 1236 | 81 | |||
| GenBank mRNA | JV567606.1 | - | 1069 | 81 | ||
More Information
Set Membership: |
|

CTSB
Genomic Map