Assay Details
| Gene Symbol: PPARA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033029.1 | NP_001028201.1 | 214 | 135 | ||
| XM_015150551.2 | XP_015006037.2 | 513 | 135 | |||
| XM_015150552.2 | XP_015006038.2 | 951 | 135 | |||
| XM_028827106.1 | XP_028682939.1 | 436 | 135 | |||
| XM_028827107.1 | XP_028682940.1 | 508 | 135 | |||
| XM_028827109.1 | XP_028682942.1 | 273 | 135 | |||
| GenBank mRNA | DQ062812.1 | - | 214 | 135 | ||
| JU476101.1 | - | 353 | 135 | |||
| JV717406.1 | - | 360 | 135 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PPARA
Genomic Map