Assay Details
| Gene Symbol: NEK7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258200.1 | NP_001245129.1 | 1124 | 73 | ||
| XM_015118870.2 | XP_014974356.1 | 871 | 73 | |||
| GenBank mRNA | JU322038.1 | - | 1078 | 73 | ||
| JU470681.1 | - | 1124 | 73 | |||
| JV046237.1 | - | 1013 | 73 | |||
More Information
Set Membership: |
|

NEK7
Genomic Map