Assay Details
| Gene Symbol: BRF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015145163.2 | XP_015000649.1 | 363 | 97 | ||
| GenBank mRNA | JV655300.1 | - | 352 | 97 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

BRF2| Gene Symbol: BRF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015145163.2 | XP_015000649.1 | 363 | 97 | ||
| GenBank mRNA | JV655300.1 | - | 352 | 97 | ||
Set Membership: |
|