Assay Details
| Gene Symbol: EXT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258133.1 | NP_001245062.1 | 2123 | 59 | ||
| GenBank mRNA | JU471485.1 | - | 2123 | 59 | ||
| JV636023.1 | - | 2101 | 59 | |||
| JV709333.1 | - | 1366 | 59 | |||
More Information
Set Membership: |
|

EXT1
Genomic Map