Assay Details
| Gene Symbol: PPM1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266202.1 | NP_001253131.1 | 1402 | 93 | ||
| XM_015112222.2 | XP_014967708.1 | 1461 | 93 | |||
| XM_015112224.2 | XP_014967710.1 | 1461 | 93 | |||
| GenBank mRNA | JU319660.1 | - | 1162 | 93 | ||
| JU319661.1 | - | 1162 | 93 | |||
| JU471866.1 | - | 1333 | 93 | |||
| JU471867.1 | - | 1333 | 93 | |||
| JV043666.1 | - | 1402 | 93 | |||
| JV043667.1 | - | 1402 | 93 | |||
| JV668882.1 | - | 1404 | 93 | |||
More Information
Set Membership: |
|

PPM1B
Genomic Map