Assay Details
| Gene Symbol: BCAS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015148747.2 | XP_015004233.2 | 2227 | 94 | ||
| XM_015148750.2 | XP_015004236.2 | 2092 | 94 | |||
| XM_015148753.2 | XP_015004239.2 | 1858 | 94 | |||
| XM_028828518.1 | XP_028684351.1 | 2050 | 94 | |||
| XM_028828519.1 | XP_028684352.1 | 2026 | 94 | |||
| XM_028828520.1 | XP_028684353.1 | 1816 | 94 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

BCAS1
Genomic Map