Assay Details
| Gene Symbol: SLC19A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015132711.2 | XP_014988197.1 | 1285 | 75 | ||
| XM_015132713.2 | XP_014988199.1 | 1638 | 75 | |||
| XM_028845237.1 | XP_028701070.1 | 1471 | 75 | |||
| GenBank mRNA | JV626462.1 | - | 1190 | 75 | ||
More Information
Set Membership: |
|

SLC19A1
Genomic Map