Assay Details
Gene Symbol: MTMR8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001097351.3 | XP_001097351.1 | 689 | 67 | ||
XM_028842498.1 | XP_028698331.1 | 689 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
