Assay Details
| Gene Symbol: HNRNPAB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257812.1 | NP_001244741.1 | 641 | 60 | ||
| XM_015141560.2 | XP_014997046.1 | 672 | 60 | |||
| GenBank mRNA | JU321547.1 | - | 641 | 60 | ||
| JU321725.1 | - | 641 | 60 | |||
| JU470979.1 | - | 569 | 60 | |||
| JU470980.1 | - | 569 | 60 | |||
| JV043817.1 | - | 511 | 60 | |||
| JV043818.1 | - | 511 | 60 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

HNRNPAB
Genomic Map