Assay Details
| Gene Symbol: KIFC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015136109.2 | XP_014991595.2 | 1107 | 60 | ||
| XM_015136110.2 | XP_014991596.2 | 1111 | 60 | |||
| XM_015136111.2 | XP_014991597.2 | 605 | 60 | |||
| XM_028847678.1 | XP_028703511.1 | 744 | 60 | |||
| GenBank mRNA | JU475522.1 | - | 568 | 60 | ||
| JV047007.1 | - | 325 | 60 | |||
| JV636143.1 | - | 377 | 60 | |||
More Information
Set Membership: |
|

KIFC1
Genomic Map