Assay Details
| Gene Symbol: SEPTIN5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015149712.2 | XP_015005198.1 | 1095 | 84 | ||
| XM_028827985.1 | XP_028683818.1 | 1181 | 84 | |||
| GenBank mRNA | JU344763.1 | - | 1027 | 84 | ||
| JV047415.1 | - | 989 | 84 | |||
| JV636227.1 | - | 1001 | 84 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SEPTIN5
Genomic Map