Assay Details
| Gene Symbol: CA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001085299.4 | XP_001085299.2 | 707 | 63 | ||
| XM_028841229.1 | XP_028697062.1 | 925 | 63 | |||
| GenBank mRNA | JV592959.1 | - | 168 | 63 | ||
| JV722864.1 | - | 113 | 63 | |||
More Information
Set Membership: |
|

CA7
Genomic Map