Assay Details
| Gene Symbol: MARCHF9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246272.1 | NP_001233201.1 | 517 | 72 | ||
| GenBank mRNA | HQ450767.1 | - | 517 | 72 | ||
| JV665735.1 | - | 468 | 72 | |||
More Information
Set Membership: |
|

MARCHF9
Genomic Map