Assay Details
| Gene Symbol: DERL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194187.2 | NP_001181116.1 | 362 | 89 | ||
| XM_015118299.2 | XP_014973785.1 | 372 | 89 | |||
| XM_028835627.1 | XP_028691460.1 | 321 | 89 | |||
| GenBank mRNA | JU320496.1 | - | 323 | 89 | ||
| JU475798.1 | - | 333 | 89 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

DERL2
Genomic Map