Assay Details
| Gene Symbol: STK38L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261495.1 | NP_001248424.1 | 921 | 73 | ||
| XM_015151154.2 | XP_015006640.1 | 978 | 73 | |||
| XM_015151156.2 | XP_015006642.1 | 1082 | 73 | |||
| XM_028828936.1 | XP_028684769.1 | 958 | 73 | |||
| GenBank mRNA | JU319754.1 | - | 905 | 73 | ||
| JU474779.1 | - | 848 | 73 | |||
| JV044856.1 | - | 921 | 73 | |||
More Information
Set Membership: |
|

STK38L
Genomic Map